Assay Details
| Gene Symbol: pip4k2ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200101.2 | NP_956395.2 | 9 - 10 | 1178 | 72 | |
| GenBank mRNA | BC045883.1 | - | 9 - 10 | 1315 | 72 | |
| BC071466.1 | - | 9 - 10 | 1178 | 72 | ||
More Information
Set Membership: |
|

pip4k2ca
Genomic Map