Assay Details
| Gene Symbol: fam49a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001324493.1 | NP_001311422.1 | 8 - 9 | 883 | 67 | |
| NM_001324495.1 | NP_001311424.1 | 8 - 9 | 887 | 67 | ||
| NM_200108.2 | NP_956402.1 | 8 - 9 | 884 | 67 | ||
| XM_005160645.4 | XP_005160702.1 | 9 - 10 | 904 | 67 | ||
| XM_005160646.4 | XP_005160703.1 | 9 - 10 | 950 | 67 | ||
| XM_005160648.4 | XP_005160705.1 | 8 - 9 | 872 | 67 | ||
| XM_009294701.3 | XP_009292976.1 | 9 - 10 | 1302 | 67 | ||
| XM_021468373.1 | XP_021324048.1 | 9 - 10 | 931 | 67 | ||
| GenBank mRNA | AY577005.1 | - | 8 - 9 | 885 | 67 | |
| BC045305.1 | - | 8 - 9 | 852 | 67 | ||
| BC065321.1 | - | 7 - 8 | 776 | 67 | ||
| EH477687.1 | - | 4 - 5 | 470 | 67 | ||
| EH541909.1 | - | 8 - 9 | 824 | 67 | ||
More Information
Set Membership: |
|

fam49a
Genomic Map