Assay Details
| Gene Symbol: elof1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200386.1 | NP_956680.1 | 2 - 3 | 203 | 124 | |
| XM_005168475.4 | XP_005168532.1 | 2 - 3 | 216 | 124 | ||
| GenBank mRNA | BC053290.1 | - | 2 - 3 | 203 | 124 | |
More Information
Set Membership: |
|

elof1
Genomic Map