Assay Details
| Gene Symbol: crispld1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200470.1 | NP_956764.1 | 7 - 8 | 1136 | 98 | |
| XM_009298567.3 | XP_009296842.1 | 7 - 8 | 1159 | 98 | ||
| GenBank mRNA | BC055176.1 | - | 7 - 8 | 1136 | 98 | |
More Information
Set Membership: |
|

crispld1b
Genomic Map