Assay Details
| Gene Symbol: crispld1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200470.1 | NP_956764.1 | 8 - 9 | 1203 | 119 | |
| GenBank mRNA | BC055176.1 | - | 8 - 9 | 1203 | 119 | |
More Information
Set Membership: |
|

crispld1b
Genomic Map