Assay Details
| Gene Symbol: onecut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200573.1 | NP_956867.1 | 1 - 2 | 1335 | 52 | |
| GenBank mRNA | AY835394.1 | - | 1 - 2 | 1109 | 52 | |
| BC056601.1 | - | 1 - 2 | 1335 | 52 | ||
More Information
Set Membership: |
|

onecut1
Genomic Map