Assay Details
| Gene Symbol: tubg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320515.1 | NP_001307444.1 | 5 - 6 | 590 | 101 | |
| NM_200908.2 | NP_957202.1 | 5 - 6 | 590 | 101 | ||
| XM_017353659.2 | XP_017209148.1 | 5 - 6 | 597 | 101 | ||
| GenBank mRNA | BC045486.1 | - | 559 | 101 | ||
| BC159223.1 | - | 5 - 6 | 584 | 101 | ||
| EH487910.1 | - | 5 - 6 | 598 | 101 | ||
More Information
Set Membership: |
|

tubg1
Genomic Map