Assay Details
| Gene Symbol: tubg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320515.1 | NP_001307444.1 | 6 - 7 | 722 | 62 | |
| NM_200908.2 | NP_957202.1 | 6 - 7 | 722 | 62 | ||
| XM_017353659.2 | XP_017209148.1 | 6 - 7 | 729 | 62 | ||
| GenBank mRNA | BC045486.1 | - | 691 | 62 | ||
| BC159223.1 | - | 6 - 7 | 716 | 62 | ||
| EH487910.1 | - | 6 - 7 | 730 | 62 | ||
More Information
Set Membership: |
|

tubg1
Genomic Map