Assay Details
| Gene Symbol: pacsin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200989.1 | NP_957283.1 | 8 - 9 | 1158 | 64 | |
| XM_005166593.3 | XP_005166650.1 | 8 - 9 | 1390 | 64 | ||
| XM_005166594.3 | XP_005166651.1 | 7 - 8 | 1322 | 64 | ||
| XM_005166595.4 | XP_005166652.1 | 7 - 8 | 1186 | 64 | ||
| XM_009303470.3 | XP_009301745.2 | 7 - 8 | 1154 | 64 | ||
| XM_021477440.1 | XP_021333115.1 | 7 - 8 | 1154 | 64 | ||
| GenBank mRNA | BC046073.1 | - | 8 - 9 | 1158 | 64 | |
More Information
Set Membership: |
|

pacsin3
Genomic Map