Assay Details
| Gene Symbol: zdhhc16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001362406.1 | NP_001349335.1 | 9 - 10 | 1111 | 86 | |
| NM_201042.2 | NP_957336.2 | 10 - 11 | 1225 | 86 | ||
| XM_009307300.3 | XP_009305575.1 | 9 - 10 | 822 | 86 | ||
| XM_017358721.2 | XP_017214210.1 | 10 - 11 | 1130 | 86 | ||
| GenBank mRNA | BC055620.1 | - | 10 - 11 | 1196 | 86 | |
More Information
Set Membership: |
|

zdhhc16a
Genomic Map