Assay Details
| Gene Symbol: zdhhc16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201042.2 | NP_957336.2 | 8 - 9 | 1018 | 123 | |
| XM_009307300.3 | XP_009305575.1 | 7 - 8 | 615 | 123 | ||
| XM_017358721.2 | XP_017214210.1 | 8 - 9 | 923 | 123 | ||
| GenBank mRNA | BC055620.1 | - | 8 - 9 | 123 | ||
More Information
Set Membership: |
|

zdhhc16a
Genomic Map