Assay Details
| Gene Symbol: ube2d1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201110.1 | NP_957404.1 | 6 - 7 | 459 | 98 | |
| GenBank mRNA | BC055599.1 | - | 6 - 7 | 459 | 98 | |
More Information
Set Membership: |
|

ube2d1a
Genomic Map