Assay Details
| Gene Symbol: arrb2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201124.1 | NP_957418.1 | 5 - 6 | 492 | 72 | |
| XM_009303039.3 | XP_009301314.1 | 4 - 5 | 577 | 72 | ||
| XM_009303040.3 | XP_009301315.1 | 4 - 5 | 588 | 72 | ||
| XM_009303041.3 | XP_009301316.1 | 5 - 6 | 557 | 72 | ||
| GenBank mRNA | BC054595.1 | - | 5 - 6 | 492 | 72 | |
| BC164508.1 | - | 5 - 6 | 387 | 72 | ||
More Information
Set Membership: |
|

arrb2b
Genomic Map