Assay Details
| Gene Symbol: arrb2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201124.1 | NP_957418.1 | 9 - 10 | 846 | 80 | |
| XM_009303039.3 | XP_009301314.1 | 8 - 9 | 931 | 80 | ||
| GenBank mRNA | BC054595.1 | - | 9 - 10 | 846 | 80 | |
| BC164508.1 | - | 9 - 10 | 741 | 80 | ||
More Information
Set Membership: |
|

arrb2b
Genomic Map