Assay Details
| Gene Symbol: arrb2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201124.1 | NP_957418.1 | 10 - 11 | 917 | 73 | |
| XM_009303039.3 | XP_009301314.1 | 9 - 10 | 1002 | 73 | ||
| GenBank mRNA | BC054595.1 | - | 10 - 11 | 917 | 73 | |
| BC164508.1 | - | 10 - 11 | 812 | 73 | ||
More Information
Set Membership: |
|

arrb2b
Genomic Map