Assay Details
| Gene Symbol: mapkapk2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201303.1 | NP_958460.1 | 1 - 2 | 578 | 107 | |
| GenBank mRNA | BC054572.1 | - | 1 - 2 | 578 | 107 | |
More Information
Set Membership: |
|

mapkapk2a
Genomic Map