Assay Details
| Gene Symbol: znf706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198889.2 | NP_001185818.1 | 2 - 3 | 555 | 75 | |
| NM_001198890.2 | NP_001185819.1 | 2 - 3 | 536 | 75 | ||
| NM_001198891.2 | NP_001185820.1 | 2 - 3 | 349 | 75 | ||
| NM_205612.3 | NP_991175.1 | 2 - 3 | 330 | 75 | ||
| GenBank mRNA | AY398376.1 | - | 2 - 3 | 261 | 75 | |
| BC076112.1 | - | 2 - 3 | 223 | 75 | ||
| BC154199.1 | - | 2 - 3 | 556 | 75 | ||
| CN179006.1 | - | 2 - 3 | 555 | 75 | ||
| CT724627.2 | - | 2 - 3 | 199 | 75 | ||
| EH439585.1 | - | 2 - 3 | 264 | 75 | ||
More Information
Set Membership: |
|

znf706
Genomic Map