Assay Details
| Gene Symbol: myoz1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205678.1 | NP_991241.1 | 4 - 5 | 610 | 71 | |
| GenBank mRNA | BC065451.1 | - | 4 - 5 | 610 | 71 | |
| BC155186.1 | - | 4 - 5 | 608 | 71 | ||
More Information
Set Membership: |
|

myoz1a
Genomic Map