Assay Details
| Gene Symbol: kat7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212635.1 | NP_997800.1 | 3 - 4 | 346 | 136 | |
| XM_021480026.1 | XP_021335701.1 | 2 - 3 | 359 | 136 | ||
| GenBank mRNA | BC045314.1 | - | 3 - 4 | 346 | 136 | |
| BC075772.1 | - | 3 - 4 | 377 | 136 | ||
More Information
Set Membership: |
|

kat7b
Genomic Map