Assay Details
| Gene Symbol: pa2g4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212641.2 | NP_997806.1 | 5 - 6 | 551 | 75 | |
| GenBank mRNA | BC056591.1 | - | 5 - 6 | 523 | 75 | |
| BC071536.1 | - | 5 - 6 | 551 | 75 | ||
More Information
Set Membership: |
|

pa2g4b
Genomic Map