Assay Details
| Gene Symbol: capza1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212697.1 | NP_997862.1 | 1 - 2 | 79 | 69 | |
| GenBank mRNA | BC047860.1 | - | 1 - 2 | 79 | 69 | |
| BC067561.1 | - | 1 - 2 | 59 | 69 | ||
More Information
Set Membership: |
|

capza1a
Genomic Map