Assay Details
| Gene Symbol: capza1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212697.1 | NP_997862.1 | 2 - 3 | 140 | 73 | |
| GenBank mRNA | BC047860.1 | - | 2 - 3 | 140 | 73 | |
| BC067561.1 | - | 2 - 3 | 120 | 73 | ||
More Information
Set Membership: |
|

capza1a
Genomic Map