Assay Details
| Gene Symbol: chst14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212829.1 | NP_997994.1 | 1 - 2 | 310 | 70 | |
| GenBank mRNA | AB089140.1 | - | 1 - 2 | 310 | 70 | |
| BC115151.1 | - | 1 - 2 | 408 | 70 | ||
More Information
Set Membership: |
|

chst14
Genomic Map