Assay Details
| Gene Symbol: nr5a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212834.1 | NP_997999.1 | 3 - 4 | 401 | 70 | |
| GenBank mRNA | AY212920.1 | - | 3 - 4 | 401 | 70 | |
| BC162252.1 | - | 3 - 4 | 299 | 70 | ||
| BC162260.1 | - | 3 - 4 | 299 | 70 | ||
More Information
Set Membership: |
|

nr5a1b
Genomic Map