Assay Details
| Gene Symbol: sdr16c5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212878.2 | NP_998043.2 | 4 - 5 | 908 | 67 | |
| GenBank mRNA | BC066732.1 | - | 4 - 5 | 821 | 67 | |
| EH548862.1 | - | 4 - 5 | 936 | 67 | ||
| EH566802.1 | - | 3 - 4 | 474 | 67 | ||
More Information
Set Membership: |
|

sdr16c5a
Genomic Map