Assay Details
| Gene Symbol: sdr16c5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212878.2 | NP_998043.2 | 5 - 6 | 1058 | 91 | |
| GenBank mRNA | BC066732.1 | - | 5 - 6 | 971 | 91 | |
| EH566802.1 | - | 2 - 3 | 324 | 91 | ||
More Information
Set Membership: |
|

sdr16c5a
Genomic Map