Assay Details
| Gene Symbol: otud3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212922.1 | NP_998087.1 | 7 - 8 | 1036 | 58 | |
| GenBank mRNA | BC067689.1 | - | 7 - 8 | 1036 | 58 | |
More Information
Set Membership: |
|

otud3
Genomic Map