Assay Details
| Gene Symbol: fbxl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212942.1 | NP_998107.1 | 3 - 4 | 726 | 84 | |
| GenBank mRNA | BC067674.1 | - | 3 - 4 | 726 | 84 | |
More Information
Set Membership: |
|

fbxl15
Genomic Map