Assay Details
| Gene Symbol: atp6ap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213023.2 | NP_998188.2 | 6 - 7 | 717 | 83 | |
| XM_009296094.3 | XP_009294369.1 | 6 - 7 | 712 | 83 | ||
| GenBank mRNA | BC059542.1 | - | 6 - 7 | 635 | 83 | |
| EH507403.1 | - | 6 - 7 | 711 | 83 | ||
More Information
Set Membership: |
|

atp6ap2
Genomic Map