Assay Details
| Gene Symbol: galnt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213196.1 | NP_998361.1 | 5 - 6 | 1190 | 62 | |
| GenBank mRNA | BC067340.1 | - | 5 - 6 | 1190 | 62 | |
More Information
Set Membership: |
|

galnt6
Genomic Map