Assay Details
| Gene Symbol: nup42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213210.1 | NP_998375.1 | 3 - 4 | 453 | 69 | |
| GenBank mRNA | BC065440.1 | - | 3 - 4 | 453 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nup42| Gene Symbol: nup42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213210.1 | NP_998375.1 | 3 - 4 | 453 | 69 | |
| GenBank mRNA | BC065440.1 | - | 3 - 4 | 453 | 69 | |
Set Membership: |
|