Assay Details
| Gene Symbol: srsf2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213382.1 | NP_998547.1 | 2 - 3 | 816 | 146 | |
| GenBank mRNA | BC046045.1 | - | 2 - 3 | 816 | 146 | |
More Information
Set Membership: |
|

srsf2a
Genomic Map