Assay Details
| Gene Symbol: stk24b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213477.1 | NP_998642.1 | 3 - 4 | 407 | 71 | |
| XM_005165833.4 | XP_005165890.1 | 3 - 4 | 989 | 71 | ||
| GenBank mRNA | BC054651.1 | - | 3 - 4 | 407 | 71 | |
More Information
Set Membership: |
|

stk24b
Genomic Map