Assay Details
| Gene Symbol: pank1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213480.2 | NP_998645.2 | 2 - 3 | 560 | 74 | |
| XM_005158505.4 | XP_005158562.1 | 2 - 3 | 920 | 74 | ||
| GenBank mRNA | BC054910.1 | - | 2 - 3 | 560 | 74 | |
More Information
Set Membership: |
|

pank1a
Genomic Map