Assay Details
| Gene Symbol: actr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213488.1 | NP_998653.1 | 8 - 9 | 1522 | 68 | |
| GenBank mRNA | BC055228.1 | - | 8 - 9 | 1522 | 68 | |
More Information
Set Membership: |
|

actr5
Genomic Map