Assay Details
| Gene Symbol: nedd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213506.1 | NP_998671.1 | 2 - 3 | 326 | 58 | |
| XM_005163004.4 | XP_005163061.1 | 2 - 3 | 359 | 58 | ||
| GenBank mRNA | BC058053.1 | - | 2 - 3 | 326 | 58 | |
More Information
Set Membership: |
|

nedd1
Genomic Map