Assay Details
| Gene Symbol: cyth1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214682.2 | NP_999847.1 | 4 - 5 | 438 | 74 | |
| XM_017353253.2 | XP_017208742.1 | 4 - 5 | 525 | 74 | ||
| XM_017353258.2 | XP_017208747.1 | 4 - 5 | 356 | 74 | ||
| GenBank mRNA | AY574952.1 | - | 74 | |||
More Information
Set Membership: |
|

cyth1a
Genomic Map