Assay Details
| Gene Symbol: gpm6ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214687.1 | NP_999852.1 | 3 - 4 | 389 | 62 | |
| XM_005159981.4 | XP_005160038.1 | 3 - 4 | 882 | 62 | ||
| XM_005159982.3 | XP_005160039.1 | 3 - 4 | 465 | 62 | ||
| GenBank mRNA | AY070264.1 | - | 3 - 4 | 389 | 62 | |
| AY070266.1 | - | 3 - 4 | 389 | 62 | ||
More Information
Set Membership: |
|

gpm6ab
Genomic Map