Assay Details
| Gene Symbol: gpm6ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214687.1 | NP_999852.1 | 6 - 7 | 704 | 57 | |
| GenBank mRNA | AY070266.1 | - | 6 - 7 | 704 | 57 | |
More Information
Set Membership: |
|

gpm6ab
Genomic Map