Assay Details
| Gene Symbol: adcyap1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214715.2 | NP_999880.1 | 4 - 5 | 683 | 79 | |
| GenBank mRNA | AF329730.1 | - | 4 - 5 | 626 | 79 | |
| BC083192.1 | - | 4 - 5 | 651 | 79 | ||
| BC164866.1 | - | 3 - 4 | 377 | 79 | ||
| EH442184.1 | - | 4 - 5 | 668 | 79 | ||
More Information
Set Membership: |
|

adcyap1b
Genomic Map