Assay Details
| Gene Symbol: scnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001813.1 | NP_001001813.1 | 3 - 4 | 294 | 75 | |
| XM_005158161.4 | XP_005158218.1 | 3 - 4 | 315 | 75 | ||
| GenBank mRNA | AY398412.1 | - | 3 - 4 | 294 | 75 | |
| BC091527.1 | - | 3 - 4 | 270 | 75 | ||
| BC110091.1 | - | 3 - 4 | 296 | 75 | ||
More Information
Set Membership: |
|

scnm1
Genomic Map