Assay Details
| Gene Symbol: ccn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001826.2 | NP_001001826.2 | 2 - 3 | 374 | 100 | |
| GenBank mRNA | BC057506.1 | - | 3 - 4 | 358 | 100 | |
| BC067668.1 | - | 2 - 3 | 374 | 100 | ||
More Information
Set Membership: |
|

ccn1
Genomic Map