Assay Details
| Gene Symbol: ccn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001826.2 | NP_001001826.2 | 3 - 4 | 731 | 122 | |
| GenBank mRNA | BC057506.1 | - | 4 - 5 | 715 | 122 | |
| BC067668.1 | - | 3 - 4 | 731 | 122 | ||
More Information
Set Membership: |
|

ccn1
Genomic Map