Assay Details
| Gene Symbol: th2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001829.1 | NP_001001829.1 | 2 - 3 | 475 | 110 | |
| XM_005164734.4 | XP_005164791.1 | 2 - 3 | 803 | 110 | ||
| GenBank mRNA | AY620823.1 | - | 2 - 3 | 475 | 110 | |
| BC162637.1 | - | 2 - 3 | 470 | 110 | ||
More Information
Set Membership: |
|

th2
Genomic Map