Assay Details
| Gene Symbol: th2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001829.1 | NP_001001829.1 | 5 - 6 | 681 | 101 | |
| GenBank mRNA | AY620823.1 | - | 5 - 6 | 681 | 101 | |
More Information
Set Membership: |
|

th2
Genomic Map