Assay Details
| Gene Symbol: fabp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002076.2 | NP_001002076.1 | 2 - 3 | 292 | 68 | |
| GenBank mRNA | BC072553.1 | - | 2 - 3 | 275 | 68 | |
| EH493621.1 | - | 2 - 3 | 325 | 68 | ||
| EH505333.1 | - | 2 - 3 | 321 | 68 | ||
More Information
Set Membership: |
|

fabp6
Genomic Map