Assay Details
| Gene Symbol: parp6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002537.1 | NP_001002537.1 | 7 - 8 | 672 | 68 | |
| XM_021467633.1 | XP_021323308.1 | 7 - 8 | 808 | 68 | ||
| GenBank mRNA | BC076271.1 | - | 7 - 8 | 672 | 68 | |
| Gene Symbol: LOC100537098 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017351978.2 | XP_017207467.2 | 720 | 68 | ||
| XM_021467719.1 | XP_021323394.1 | 720 | 68 | |||
More Information
Set Membership: |
|

parp6a
Genomic Map