Assay Details
| Gene Symbol: preb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002638.1 | NP_001002638.1 | 9 - 10 | 1340 | 133 | |
| GenBank mRNA | BC075936.1 | - | 9 - 10 | 1340 | 133 | |
More Information
Set Membership: |
|

preb
Genomic Map