Assay Details
| Gene Symbol: rpl29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003434.2 | NP_001003434.1 | 3 - 4 | 181 | 102 | |
| NM_001243178.1 | NP_001230107.1 | 3 - 4 | 184 | 102 | ||
| GenBank mRNA | BC076328.1 | - | 3 - 4 | 147 | 102 | |
| EH502517.1 | - | 3 - 4 | 171 | 102 | ||
| EH541004.1 | - | 3 - 4 | 210 | 102 | ||
| EH543902.1 | - | 3 - 4 | 187 | 102 | ||
| EH564579.1 | - | 1 - 2 | 176 | 102 | ||
More Information
Set Membership: |
|

rpl29
Genomic Map