Assay Details
| Gene Symbol: capn2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003485.2 | NP_001003485.1 | 14 - 15 | 1665 | 65 | |
| GenBank mRNA | BC076557.1 | - | 14 - 15 | 1646 | 65 | |
| BC154289.1 | - | 14 - 15 | 1645 | 65 | ||
More Information
Set Membership: |
|

capn2l
Genomic Map